Kininogen-Based Thrombin Inhibitor
4025629.0025 4025629.0005
H-2282.0025BAEA 1784.3 USD
H-2282.0025BA H-2282.0005BA
Kininogen-Based Thrombin Inhibitor
See also lepirudin (H-6618) and bivalirudin (H-6668).
The sequence FQVVCG-amide is derived from the thiol-protease inhibitory sequence of kininogens.