Histogranin
H-2266.0500 H-2266.1000
H-2266.0500BAEA 489.01 USD
H-2266.0500BA H-2266.1000BA
Histogranin
The neuroactive peptide histogranin was originally isolated from bovine adrenal medulla. It is an NMDA receptor antagonist that acts like phencyclidine. Injection of the peptide (5-100nmol) in mice produced a dosedependent protection against NMDA-induced convulsions.