Moth Cytochrome C (MCC) Fragment
Supplier: Bachem
4025851.0005 4025851.0001
H-2252.0001BAEA 222.82 USD
H-2252.0001BA H-2252.0005BA
Moth Cytochrome C (MCC) Fragment
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
MCC (88-103), ANERADLIAYLQQATK, is an antigenic peptide that prevents the development of mature 2B4 TCRhi CD4⁺8⁺ thymocytes. This analog, containing Gln at residue 99 instead of Lys, competitively blocks antigen induced IL-2 production by the 2B4 hybridoma and it is therefore suggested that it is acting as a competitive antagonist.