TRAP-7 Trifluoroacetate, Bachem
Supplier: BACHEM AMERICAS INC
4025850.0005
H-2234.0005BAEA 641.65 USD
H-2234.0005BA
TRAP-7 Trifluoroacetate, Bachem
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
PAR-1 peptides are also called TRAP peptides (thrombin receptor-activating peptides).
The thrombin receptor agonist TRAP-7, SFLLRNP, corresponds to residues 42-48 of the thrombin receptor. TRAP-7 is as effective as thrombin in inducing Ca²⁺ mobilization and protein kinase C activation in some T cell lines.