Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcineurin Substrate

Supplier: BACHEM AMERICAS INC
H-2084.0001 H-2084.0005
Sequence: H-Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser-Val-Ala-Ala-Glu-OH
CAS-No.: 113873-67-9
Mol.weight: 2112.37
SumFormula: C₉₂H₁₅₀N₂₈O₂₉
Bulk item no.: H-2084
Synonym(s):
EINECS no.: -
MDL no.: MFCD18782129

The 19-peptide DLDVPIPGRFDRRVSVAAE is a fragment of the RII subunit of the cAMP-dependent protein kinase. It is successfully used in a nonradioactive assay in which both phosphorylation by protein kinase and dephosphorylation by protein phosphatase 2B (calcineurin) are monitored by the simultaneous quantitative determination of phosphorylated and non-phosphorylated peptide using HPLC on an RP18 column with UV detection. This method allows the measurement of enzyme kinetics as well as the characterization of novel inhibitors.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-2084.0001 H-2084.0001BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

5 mg H-2084.0005 H-2084.0005BA Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

CAS No. 113873-67-9
Molecular Weight 2112.37
Molecular Formula C₉₂H₁₅₀N₂₈O₂₉
Storage Temperature -20 ± 5 °C
Salt Trifluoroacetate
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty