Fibronectin (196-203)
Supplier: Bachem
4030611.0005 4030611.0025
H-2064.0005BAEA 420.99 USD
H-2064.0005BA H-2064.0025BA
Fibronectin (196-203)
The fragment SRNRCNDQ-amide, a sequence derived from the amino-terminal heparinbinding domain of fibronectin, is a potent stimulator of tissue plasminogen activator (t-PA)-catalyzed plasminogen activation. The peptide induces a 15-fold increase in the kcat/Km of the activation reaction. However it influences neither the t-PA nor the plasmin amidolytic activity.