Ac-Arg-Phe-Met-Trp-Met-Lys-NH₂
H-1994.0005 H-1994.0025
H-1994.0025BAEA 651.65 USD
H-1994.0025BA H-1994.0005BA
Ac-Arg-Phe-Met-Trp-Met-Lys-NH₂
Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.
The acetalins are opioid receptor ligands with high affinity for µ and K₃ receptors, weak affinity for K₁ receptors and no affinity for K₂ receptors.
Ac-RFMWMK-amide.