alpha-Substance IB
Supplier: Bachem
4002167.0100 4002167.0025
H-1840.0025BAEA 669.48 USD
H-1840.0025BA H-1840.0100BA
alpha-Substance IB
α-Substance IB is produced by α-mating type cells of Saccharomyces cerevisiae and induces sexual agglutinability in the opposite a type cells. RGPFPI corresponds to β-casein (200-205), it has been detected in casein hydrolysates. The hexapeptide has also found use as test compound in the development and evaluation of analytical methods for peptides. CAS Number (acetate) 64566-61-6.