(Sar¹,Thr⁸)-Angiotensin II
4000892.0025
H-1745.0025BAEA 275.57 USD
H-1745.0025BA
(Sar¹,Thr⁸)-Angiotensin II
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
Specific angiotensin II competitive antagonist.