Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

rec IL-1β (murine)

Supplier: BACHEM AMERICAS INC
Sequence:
CAS-No.:
Mol.weight:
SumFormula:
Bulk item no.: H-1736
Synonym(s): rec Interleukin-1β (murine)
EINECS no.:
MDL no.: MFCD00163057

Interleukins are soluble proteins that act as local hormones. They possess powerful immunoregulatory properties.

Murine IL-1β is a 15-17 kD polypeptide that shares only 26% amino acid sequence homology with IL-1α. They are however structurally related and share therefore the same receptor and have essentially identical functions, including prostaglandin synthesis, activation of T-lymphocytes, regulation of growth and function of B-lymphocytes and a variety of non-immune related functions.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 µg H-1736.0010 H-1736.0010BA Each (10µG) Retrieving Restricted
2 µg H-1736.0002 H-1736.0002BA Each (2µG) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name IL1B
Synonyms rec Interleukin-1b (murine)
Protein/Peptide Type Recombinant
Species Mouse
Storage Conditions −20 ± 5 °C
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty