Human Recombinant Neurotrophin 3
Supplier: Bachem
4031114.0002
H-1726.0002BAEA 136.22 USD
H-1726.0002BA
Human Recombinant Neurotrophin 3
Please see also H-2512 and G-2080.
Human NT-3 is a member of the family of neurotrophic factors that control the development, survival and phenotypic differentiation of nerve and/or glial cells.