Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Angiotensin I/II (1-7)

Supplier: BACHEM AMERICAS INC
H-1715.0025 H-1715.0100
Sequence: H-Asp-Arg-Val-Tyr-Ile-His-Pro-OH
CAS-No.: 51833-78-4
Mol.weight: 899.02
SumFormula: C₄₁H₆₂N₁₂O₁₁
Bulk item no.: H-1715
Synonym(s):
EINECS no.: -
MDL no.: MFCD00153513

The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.

Angiotensin (1-7), DRVYIHP, an enzymatically produced peptide of the renin-angiotensin system, shows pharmacologically distinct, tissue-specific actions that differ from those of angiotensin II. Angiotensin (1-7) shows considerable cardiovascular and antiangiogenic activity. Iusuf et al. reviewed its phamacological properties and pharmacotherapeutic perspectives. Moreover, the heptapeptide was shown to inhibit the growth of human lung cancer cells in vitro and to reduce the size of human lung cancer xenografts in vivo.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 mg H-1715.0100 H-1715.0100BA Each (100mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

25 mg H-1715.0025 H-1715.0025BA Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Angiotensin I/II (1-7)
CAS No. 51833-78-4
Molecular Weight 899.02
Molecular Formula C₄₁H₆₂N₁₂O₁₁
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty