rec RANTES (human)
H-1706.0020 H-1706.0005
H-1706.0020BAEA 315.31 USD
H-1706.0020BA H-1706.0005BA
rec RANTES (human)
The highly basic 8kD polypeptide, RANTES, is a chemoattractant for monocytes but not neutrophils in vitro. It also displays chemoattractant activity on specific T cell subsets (CD4+/CD45RO+).