Bifunctional Antiplatelet Agent
Supplier: Bachem
4020279.0001 4020279.0005
H-1662.0005BAEA 1089.64 USD
H-1662.0005BA H-1662.0001BA
Bifunctional Antiplatelet Agent
Bifunctional antiplatelet agent blocked the binding of ¹²⁵I-fibrinogen to ADP-stimulated platelets with an IC₅₀ of 1.2 µM and inhibited ADP-induced platelet aggregation with an IC₅₀ of 29 µM. It was shown to be bifunctional in that it not only inhibited the binding of fibrinogen to glycoprotein IIb/IIIa, but also inhibited the binding of von Willebrand factor to glycoprotein Ib.