HSV-1 Glycoprotein (gB) (497-507)
Supplier: Bachem
4025035.0001 4025035.0005
H-1628.0001BAEA 334.74 USD
H-1628.0001BA H-1628.0005BA
HSV-1 Glycoprotein (gB) (497-507)
The undecapeptide TSSIEFARLQF represents a H-2Kb T cell epitope of HSV-1 gB. It activates CD8+ cytotoxic T lymphocytes in vivo in a manner independent of CD4+ T cells.