Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Prion Protein (106-126) (human)

Supplier: BACHEM AMERICAS INC
Sequence: H-Lys-Thr-Asn-Met-Lys-His-Met-Ala-Gly-Ala-Ala-Ala-Ala-Gly-Ala-Val-Val-Gly-Gly-Leu-Gly-OH
CAS-No.: 148439-49-0
Mol.weight: 1912.27
SumFormula: C₈₀H₁₃₈N₂₆O₂₄S₂
Bulk item no.: H-1566
Synonym(s): Prion Protein Fragment (106-126), human
EINECS no.: -
MDL no.: MFCD18782118

For additional products in the field of Alzheimer's disease, please see Amyloid Peptides, β-Secretase Inhibitors and Substrates and Presenilin-1 (331-349)-Cys (H-3988).

An altered form of the large cellular prion protein accumulates in the CNS of patients with neurodegenerative disorders, and its protease-resistant core aggregates extracellularly into amyloid fibrils. This process is accompanied by nerve cell loss. The 21-peptide KTNMKHMAGAAAAGAVVGGLG is used as a model to investigate neurodegeneration in prion diseases. Prion Protein (106-126) forms fibrils in vitro and causes apoptotic cell death in neuron culture. Its neurotoxicity depends on the presence of microglia and is induced by microglia. It could be demonstrated that KTNMKHMAGAAAAGAVVGGLG is able to bind to DNA, a circumstance which led to the assumption that the integral protein could also bind to DNA under appropriate solution conditions.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-1566.0500 H-1566.0500BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg H-1566.1000 H-1566.1000BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

CAS No. 148439-49-0
Molecular Weight 1912.27
Storage Temperature -20 ± 5 °C
Molecular Formula C₈₀H₁₃₈N₂₆O₂₄S₂
Salt Trifluoroacetate
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty