Ac-Met-Asp-Lys-Val-Leu-Asn-Arg-Glu-OH
Supplier: Bachem
4009000.0005
H-1440.0005BAEA 387.66 USD
H-1440.0005BA
Ac-Met-Asp-Lys-Val-Leu-Asn-Arg-Glu-OH
Proteins and Peptides
The sequence MDKVLNRE corresponds to the N-terminal octapeptide of the large T antigen of simian virus 40 (SV40) and BK polyoma virus.