Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C-Reactive Protein (CRP) (201-206)

Supplier: BACHEM AMERICAS INC
Sequence: H-Lys-Pro-Gln-Leu-Trp-Pro-OH
CAS-No.: 130348-99-1
Mol.weight: 767.93
SumFormula: C₃₈H₅₇N₉O₈
Bulk item no.: H-1438
Synonym(s):
EINECS no.: -
MDL no.: MFCD00236936

These synthetic oligopeptides correspond to a region of high amino-acid sequence similarity between uteroglobin and lipocortin I. They possess a potent anti-inflammatory activity in vivo and are strong inhibitors of phospholipase A₂ (PLA₂) in vitro. The name 'antiflammins' is proposed for these peptides, which are a valuable model for the development of novel anti-inflammatory agents of therapeutic importance. See also Cortistatin-29, H-6458, and Thymic Factor, H-4865.

CRP (201-206) inhibits neutrophil adhesion to endothelial cells and platelets through CD32.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 mg H-1438.0025 H-1438.0025BA Each (25mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

5 mg H-1438.0005 H-1438.0005BA Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

CAS No. 130348-99-1
Molecular Weight 767.93
Storage Temperature -20 ± 5 °C
Molecular Formula C₃₈H₅₇N₉O₈
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty