(Tyr⁶⁹,Ala⁷¹·⁷²,Lys⁷⁴)-C3a (69-77)
4018154.0005 4018154.0025
H-1432.0025BAEA 1685.25 USD
H-1432.0025BA H-1432.0005BA
(Tyr⁶⁹,Ala⁷¹·⁷²,Lys⁷⁴)-C3a (69-77)
The peptide YAAALKLAR, which has been modeled after the C-terminus of the anaphylatoxin C3a, constitutes a potent inhibitor of C3a activity. The Lys⁷⁴ residue contributes to the separation between deactivation and activation of the C3a receptor.