Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HIV-1 env Protein gp120 (278-292) (strains BH10, BH8, HXB2, HXB3, PV22)

Supplier: BACHEM AMERICAS INC
Sequence: H-Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys-OH
CAS-No.: 114991-28-5
Mol.weight: 1655.97
SumFormula: C₇₃H₁₂₆N₂₆O₁₈
Bulk item no.: H-1402
Synonym(s): P18IIIB#HIV gp120 Fragment (315-329)#HIV-1 env Protein gp120 (283-297) (strain BRU)#R15K, HIV-1 Inhibitory Peptide
EINECS no.: -
MDL no.: MFCD00235749

P18IIIB, derived from the V3 loop of HIV-1 IIIB gp120 efficiently blocked HIV-1 IIIB infection of several T-cell lines and of normal human T cells. RIQRGPGRAFVTIGK also blocked syncytium formation in human cells. Moreover, P18IIIB was capable of inducing HIV-1 specific cytotoxic T lymphocyte responses that could effectively kill virus-infected cells and could thus find several therapeutic applications.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-1402.0001 H-1402.0001BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

5 mg H-1402.0005 H-1402.0005BA Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

CAS No. 114991-28-5
Molecular Weight 1655.97
Storage Temperature -20 ± 5 °C
Molecular Formula C₇₃H₁₂₆N₂₆O₁₈
Synonyms HIV-1 env Protein gp120 (283-297) (strain BRU), P18IIIB, HIV gp120 Fragment (315-329)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty