Glycoprotein IIb Fragment (300-312)
Supplier: Bachem
4030668.0005 4030668.0001
H-1366.0005BAEA 1337.29 USD
H-1366.0005BA H-1366.0001BA
Glycoprotein IIb Fragment (300-312)
For HIV and SIV glycoprotein fragments please see HIV Peptides.
This GPII fragment has been shown to inhibit aggregation of washed human platelets and to inhibit the adhesion of stimulated platelets to fibrinogen, fibronectin, vitronectin and von Willebrand factor.