HIV (gp120) Fragment (421-438) Trifluoroacetate
Supplier: Bachem
4018349.1000 4018349.0500
H-1354.0500BAEA 247.64 USD
H-1354.0500BA H-1354.1000BA
HIV (gp120) Fragment (421-438) Trifluoroacetate
The octadecapeptide KQFINMWQEVGKAMYAPP derived from the CD4 attachment region of HIV gp120 inhibited the syncytial formation in vitro. According to immunological studies, the HIV (gp120) fragment 421-438 generated high titres of anti-gp120 antibodies following immunization in mice, rabbits, and goats.