H-D-Ala-D-Ala-D-Ala-D-Ala-OH
Supplier: Bachem
4008059.0250
H-1265.0250BAEA 548.65 USD
H-1265.0250BA
H-D-Ala-D-Ala-D-Ala-D-Ala-OH
Proteins and Peptides
Oligomers of L- and/or D-alanine, the trimers to the hexamers. Bachem also offers terminally modified oligoalanines as H-1005, Ac-Ala₄-OH. and H-1010, Ac-Ala₄-OMe.
Tetra-D-alanine, substrate for D-aminopeptidase from Ochrobactrum anthropi.