(Sar¹,Val⁵,Ala⁸)-Angiotensin II Trifluoroacetate, Bachem
4018035.0100 4018035.0025
H-1232.0100BAEA 826.74 USD
H-1232.0100BA H-1232.0025BA
(Sar¹,Val⁵,Ala⁸)-Angiotensin II Trifluoroacetate, Bachem
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
As a noncompetitive angiotensin II antagonist, saralasin inhibits the pressor effects of angiotensin II in rats and shows blood pressure lowering activity in humans. It is used as a diagnostic for renin-dependent hypertension.