|
![]() |
ORDER
SPECIFICATIONS
Protein/Peptide Name | Tachyplesin I | |
CAS No. | 118231-04-2 | |
Molecular Weight | 2263.78 | |
Molecular Formula | C₉₉H₁₅₁N₃₅O₁₉S₄ | |
Storage Conditions | -20 ± 5 °C | |
Salt | Trifluoroacetate |
![]() |
Processing your request... |
Are you sure you wish to return? |
||
|
Item(s) added to your Shopping Basket
|
||||||||
---|---|---|---|---|---|---|---|---|
|
With 160 years of experience in supporting science, our people have the practical knowledge and creative expertise to think big, deliver fresh ideas, and develop innovative services to help you solve your most critical business challenges.
Making lab and production operations more efficient and competitive
Offering a wide range of support and services
Lending expertise to improve quality and boost scientific productivity
Helping achieve innovation faster
Over 1,200 VWRCATALYST associates are working worldwide today at industry-leading pharmaceutical, biotech, healthcare, education, industrial, and high-tech production institutions.
For more information on VWRCATALYST, call 1.888.793.2300 or email us at vwrcatalyst@vwr.com.
VWR enables the advancement of science by providing high-quality chemicals and services, customized to your product or manufacturing needs.
We use operational excellence to deliver solutions that enable research, testing, production, and commercialization across the globe.
Our Core Capabilities Include:
Contact VWR Custom Manufacturing Services at 1.800.932.5000 or VWRCustom@vwr.com.
Email: technicalproductSupportNA@vwr.com
Phone: 1-888-VWR-LINE (1-888-897-5463)
Phone: 1-800-932-5000
Online: Contact Us
Phone: 1-888-320-4357
Online: Help Center
Online: Contact Local Team
Online: Return Request Form
![]() |
|
|
Avantor is a leading global provider of integrated, tailored solutions for the life sciences and advanced technology industries. Strengthened by the recent acquisition of VWR, the Company is a trusted partner to customers and suppliers from discovery to delivery. Collectively, we set science in motion to create a better world.