Ac-Ala-Ala-Ala-Ala-OMe
Supplier: Bachem
4010707.0250
H-1010.0250BAEA 468.47 USD
H-1010.0250BA
Ac-Ala-Ala-Ala-Ala-OMe
Proteins and Peptides
Substrate for the esterolytic activity of elastase, as the enzyme cleaves the methyl ester of Ac-Ala4-OMe much more rapidly than an amide bond.