Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Anserine · nitrate

Supplier: BACHEM AMERICAS INC

G-4555.0250 G-4555.0100
G-4555.0250BAEA 367.04 USD InStock
G-4555.0250BA G-4555.0100BA
L-Anserine · nitrate
Proteins and Peptides
Sequence: H-β-Ala-His(3-Me)-OH · nitrate
CAS-No.: 10030-52-1
Mol.weight: 303.28
SumFormula: C₁₀H₁₆N₄O₃ · HNO₃
Bulk item no.: G-4555
Synonym(s):
EINECS no.: 233-079-7
MDL no.: MFCD00037001

Like L-carnosine (G-1250), L-anserine is a histidine-containing dipeptide found in vertebrate skeletal muscles. Both are potent antioxidants with membrane-stabilizing function.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty