Gly-Gly-OH,Biotin
Supplier: Bachem
4026362.1000 4026362.0250
G-4485.0250BAEA 420.99 USD
G-4485.0250BA G-4485.1000BA
Gly-Gly-OH,Biotin
Building block for introducing biotin with a polar spacer moiety or for synthesizing biotinylated Gly-Gly-containing peptides.