H-Trp-Lys-OH

Supplier: BACHEM AMERICAS INC

4002439.0250 4002439.1000
G-3800.0250BAEA 537.12 USD InStock
G-3800.0250BA G-3800.1000BA
H-Trp-Lys-OH
Proteins and Peptides
Sequence: H-Trp-Lys-OH
CAS-No.: 51790-14-8
Mol.weight: 332.40
SumFormula: C₁₇H₂₄N₄O₃
Bulk item no.: G-3800
Synonym(s):
EINECS no.: -
MDL no.: MFCD00238201
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR