beta-Casomorphin (1-2)
Supplier: Bachem
4002382.0250
G-3625.0250BAEA 518.16 USD
G-3625.0250BA
beta-Casomorphin (1-2)
Proteins and Peptides
Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.
  • Also called β-casein exorphins.
The antihypertensive peptide isolated from fermented milk had a low inhibitory activity against angiotensin I-converting enzyme (ACE), IC₅ ₀ 720 μM. G-3625 is a possible biosynthetic precursor of endomorphin.