Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Trp-Trp-OH

Supplier: BACHEM AMERICAS INC

G-3370.0005 G-3370.0001
G-3370.0005BAEA 1910.95 USD InStock
G-3370.0005BA G-3370.0001BA
H-Trp-Trp-OH
Proteins and Peptides
Sequence: H-Trp-Trp-OH
CAS-No.: 20696-60-0
Mol.weight: 390.44
SumFormula: C₂₂H₂₂N₄O₃
Bulk item no.: G-3370
Synonym(s):
EINECS no.: -
MDL no.: MFCD00037959

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty