Human Dynorphin B (1-9)
Supplier: Bachem
4039876.0001
H-5738.0001BAEA 206.68 USD
H-5738.0001BA
Human Dynorphin B (1-9)
Proteins and Peptides
The N-terminus of dynorphin, YGGFL, corresponds to Leu-enkephalin. So, dynorphin A (1-5) amide is filed as Leu-enkephalin amide (H-2745). For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.