(D-Tyr⁵,D-Ser(tBu)⁶,Azagly¹⁰)-LHRH
Supplier: BACHEM AMERICAS INC
4035302.0005 4035302.0001
H-5734.0001BAEA 518.16 USD
H-5734.0001BA H-5734.0005BA
(D-Tyr⁵,D-Ser(tBu)⁶,Azagly¹⁰)-LHRH
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Impurity F of the Goserelin Ph. Eur. monograph.