Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Liver Cell Growth Factor

Supplier: BACHEM AMERICAS INC

H-3510.1000 H-3510.0250
H-3510.1000BAEA 358.08 USD InStock
H-3510.1000BA H-3510.0250BA
Liver Cell Growth Factor
Proteins and Peptides
Sequence: H-Gly-His-Lys-OH
CAS-No.: 72957-37-0
Mol.weight: 340.38
SumFormula: C₁₄H₂₄N₆O₄
Bulk item no.: H-3510
Synonym(s): GHK#Prezatide
EINECS no.: 277-125-4
MDL no.: MFCD00012699

GHK possesses a high affinity for copper(II) ions, with which it spontaneously forms a complex (GHK-Cu). This growth-modulating plasma tripeptide, when added at nanomolar concentrations to a wide group of cultured systems, produces a disparate set of responses ranging from the stimulation of growth and differentiation to outright toxicity. It may function by facilitating copper uptake into cells. GHK-Cu accelerates wound healing. CAS Number (net): 49557-75-7.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty