Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Gln⁵³)-Connexin 37 (51-58) (human, mouse, rat)

Supplier: BACHEM AMERICAS INC
H-3458.0001 H-3458.0005
Sequence: H-Phe-Glu-Gln-Asn-Thr-Ala-Gln-Pro-OH
CAS-No.: 155893-22-4
Mol.weight: 933.97
SumFormula: C₄₀H₅₉N₁₁O₁₅
Bulk item no.: H-3458
Synonym(s): MUT 1 (human, mouse, rat)#(Gln⁵³)-Gap Junction α-4 Protein (51-58) (human, mouse, rat)#(Gln⁵³)-Cx37 (51-58) (human, mouse, rat)
EINECS no.: -
MDL no.: MFCD00798723

MUT 1, a tumor-associated antigen peptide originally isolated from the mouse Lewis lung carcinoma, appears to be a fragment of the gap junction protein connexin 37, mutated at position 54. MUT 1 induces effective antitumor cytotoxic T lymphocytes and mediates protection against metastasis, as well as rejection of established metastasis. This is the first description of peptide vaccine efficacity in treatment of residual tumors.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-3458.0001 H-3458.0001BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

5 mg H-3458.0005 H-3458.0005BA Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name (Gln⁵³)-Connexin 37 (51-58)
Protein Synonyms MUT 1 (human, mouse, rat), (Gln53)-Gap Junction a-4 Protein (51-58) (human, mouse, rat), (Gln53)-Cx37 (51-58) (human, mouse, rat)
Species Rat, Human, Mouse Human, Mouse, Rat
CAS No. 155893-22-4
Molecular Weight 933.97
Molecular Formula C₄₀H₅₉N₁₁O₁₅
Storage Temperature -20 ± 5 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty