Ac-Pen-Arg-Gly-Asp-Cys-OH
Supplier: Bachem
4018743.0001
H-1614.0001BAEA 206.68 USD
H-1614.0001BA
Ac-Pen-Arg-Gly-Asp-Cys-OH
Thie cyclic RGD-containing pentapeptide Ac-PenRGDC has been demonstrated to be a potent inhibitor of blood platelet aggregation (IC₅₀ = 0.55 µM).