H-Gly-Phe-Arg-Gly-Asp-Gly-Gln-OH
Supplier: Bachem
4025013.0005 4025013.0001
H-1606.0001BAEA 273.66 USD
H-1606.0001BA H-1606.0005BA
H-Gly-Phe-Arg-Gly-Asp-Gly-Gln-OH
The peptide GFRGDGQ comprising the RGD-containing sequence within the basement membrane glycoprotein entactin promoted mouse trophoblast outgrowth when immobilized on the substratum, indicating that this region of the protein possesses the essential structural requirements for directing trophoblast adhesive activity.