Human Seminal Plasma Inhibin (67-94) (from Plasma (seminal))
Supplier: Bachem
4030771.1000 4030771.0500
H-1602.0500BAEA 309.56 USD
H-1602.0500BA H-1602.1000BA
Human Seminal Plasma Inhibin (67-94) (from Plasma (seminal))
This cyclic peptide suppresses significantly the circulating levels of follicle-stimulating hormone (FSH) in vivo in adult male rats. Thus this peptide, representing the carboxy-terminal region of HSPI, has inhibin-like activity and perhaps bears the active core of the protein.