H-Ala-Pro-Gly-OH, Bachem
Supplier: Bachem
4005068.0001
H-1600.0001BAEA 468.47 USD
H-1600.0001BA
H-Ala-Pro-Gly-OH, Bachem
See also H-3974, H-Lys(4-nitro-Z)-pyrrolidide · HCl (N-1710) and H-Phe-pyrrolidide (E-3275).
Inhibitor of dipeptidylpeptidase IV (DPP IV).