Procollagen Type I (212-216)
Supplier: BACHEM AMERICAS INC
4025012.0025 4025012.0005
H-1592.0025BAEA 501.51 USD
H-1592.0025BA H-1592.0005BA
Procollagen Type I (212-216)
Proteins and Peptides
A subfragment of the carboxy-terminal propeptide of type I collagen (residues 197-241) was shown to stimulate extracellular matrix biosynthesis in fibroblasts. The pentapeptide KTTKS has been found to be the minimum sequence necessary for this potent stimulation of collagen and fibronectin production in a variety of mesenchymal cells.