Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CDR-H3/C2

Supplier: BACHEM AMERICAS INC
H-1588.0500 H-1588.1000
Sequence: H-Cys-Asp-Leu-Ile-Tyr-Tyr-Asp-Tyr-Glu-Glu-Asp-Tyr-Tyr-Phe-Asp-Cys-OH
CAS-No.: 149475-21-8
Mol.weight: 2130.25
SumFormula: C₉₈H₁₂₀N₁₆O₃₄S₂
Bulk item no.: H-1588
Synonym(s):
EINECS no.: -
MDL no.: MFCD00237375

CDR-H3/C2, complementarity-determining region peptide derived from the third heavy-chain domain (CDR/H3) of the mouse monoclonal antibody F58, acts as a miniantibody and inhibited HIV-1 replication as well as syncytium formation by infected cells. Therefore, CDR-H3/C2 might be a potential tool for the treatment of HIV-infected persons.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-1588.0500 H-1588.0500BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

1 mg H-1588.1000 H-1588.1000BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name CDR-H3/C2
CAS No. 149475-21-8
Molecular Weight 2130.25
Molecular Formula C₉₈H₁₂₀N₁₆O₃₄S₂
Storage Conditions -20 ± 5 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping