Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Des-Gly)-Glutathione (reduced)

Supplier: BACHEM AMERICAS INC

G-4305.1000 G-4305.0250
G-4305.1000BAEA 1254.48 USD InStock
G-4305.1000BA G-4305.0250BA
(Des-Gly)-Glutathione (reduced)
Proteins and Peptides
Sequence: H-Glu(Cys-OH)-OH
CAS-No.: 636-58-8
Mol.weight: 250.28
SumFormula: C₈H₁₄N₂O₅S
Bulk item no.: G-4305
Synonym(s): H-γ-Glu-Cys-OH ammonium salt
EINECS no.: -
MDL no.: MFCD18782106

Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty