H-Asp-β-Ala-OH

Supplier: BACHEM AMERICAS INC

4002812.0250 4002812.1000
G-1555.0250BAEA 191.21 USD InStock
G-1555.0250BA G-1555.1000BA
H-Asp-β-Ala-OH
Proteins and Peptides
Sequence: H-Asp-β-Ala-OH
CAS-No.: 89156-89-8
Mol.weight: 204.18
SumFormula: C₇H₁₂N₂O₅
Bulk item no.: G-1555
Synonym(s):
EINECS no.: -
MDL no.: MFCD00237819
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR