Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Ala-Ala-NH₂ · HCl

Supplier: BACHEM AMERICAS INC

G-1155.0250 G-1155.1000
G-1155.0250BAEA 191.21 USD InStock
G-1155.0250BA G-1155.1000BA
H-Ala-Ala-NH₂ · HCl
Proteins and Peptides
Sequence: H-Ala-Ala-NH₂ · HCl
CAS-No.: 41036-33-3
Mol.weight: 195.65
SumFormula: C₆H₁₃N₃O₂ · HCl
Bulk item no.: G-1155
Synonym(s):
EINECS no.: -
MDL no.: MFCD00039089

Substrate for dipeptidyl aminopeptidase I.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty