High Capacity Force Gauge, Extech
Supplier: Extech
475044 475044-NIST
97049-992EA 903.17 USD
97049-992 10171-882
High Capacity Force Gauge, Extech
Dynamometers
High capacity force gauge with Push/Pull measurements in kg, lbs, oz and Newton units
The unit has 5 digit 0.4" (10mm) LCD with reversible display feature to match viewing angle. Also offers over-range, low battery, and advanced function indication.
Certifications: CE marked.
Ordering information: 1-Year Warranty. Complete with hook adaptor for tension, compression adaptors (flat, cone and chisel head), 5" extension rod, 6 AA batteries and carrying case.