Terrific Broth, Ready-Made Powder, Ultrapure, Thermo Scientific

Supplier: Thermo Fisher Scientific

J75856-A1
101174-122EA 150.37 USD
101174-122
Terrific Broth, Ready-Made Powder, Ultrapure, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
Provides greater yields of plasmid DNA and recombinant proteins than LB media by allowing for greater cell density.

Contents:
Casein Peptone: 12 g/L
Yeast Extract: 24 g/L
Potassium Phosphate Dibasic: 12.5 g/L
Potassium Phosphate Monobasic: 2.3 g/L

To use:
Add 50.8 gm powder, 4mL glycerol and 996 mL water to make 1 liter medium. Dissolve and autoclave.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR