Pocket Colorimeter™ II, Chlorine (Free), Kit with SwifTest™ DPD Reagent Dispenser, Hach
5870023
CA11014-570
Pocket Colorimeter™ II, Chlorine (Free), Kit with SwifTest™ DPD Reagent Dispenser, Hach
The new Pocket Colorimeter™ II Filter Photometer is a true go-anywhere instrument.
  • Single Parameter go anywhere portable photometer
  • USEPA approved for drinking water and wastewater
  • FactoryProgrammed
  • Waterproof, Lightweight, Rugged
  • LCD, Backlit Display, Data Logging, Recall
It's lightweight and battery operated, suitable for extended field work or quick, on-the-spot process monitoring. The instrument has two channels in which measurements can be made. Each channel will accept a user-defined calibration curve. Up to 10 standards can be used to determine the calibration. The curve is generated by a point-to-point straight line segment between each standard used. Linear and non-linear positive or negative slope calibrations can be performed. A calibration curve may also be manually entered from the keypad if a previously determined curve has been made on the Colorimeter. At least two data pairs (concentration and absorbance) are required.
Ordering information: 2 yr warranty.