Would you like to log in again?
NitraVer® 5 Nitrate Reagent Powder Pillows, Hach
Supplier: Hach
1403428 1403499 14034-99
10765-198PK 317.09 USD InStock
10765-198 97026-580 CA14034-99
NitraVer® 5 Nitrate Reagent Powder Pillows, Hach
Environmental Test Kits
For mid-range and high range nitrate determination by the NitraVer® 5 cadmium reduction method
Ordering information: Pack of 100 powder pillows.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty