AutoCAT 9000 Chlorine Amperometric Titrator, Hach
Supplier: Hach
5008100 5008100
97027-098EA 4392.13 USD
CA97027-098 97027-098
AutoCAT 9000 Chlorine Amperometric Titrator, Hach
Automatic calculation of analyte concentration
-Pre-Programmed for Easier Use
-USEPA-Approved Methods
-Automatic Calculations
-Better Control of Titration Results
Automatic hands-free operation. Easy-to-use interface. Real-time graphics and graphic print output. Automatic archiving of results. Titrant calibration. Electrode cleaning. Greater accuracy and precision than manual titrations. Real-time graphics, menu driven, LCD display.