Total Organic Carbon (TOC) Reagent Set, LR, Hach
Supplier: Hach
27603-45 2760345
89231-426EA 649.86 USD
CA27603-45 89231-426
Total Organic Carbon (TOC) Reagent Set, LR, Hach
Environmental Test Kits
For determination of Total Organic Carbon (TOC) by the Low Range Test 'N Tube™ method.
  • Affordable
  • Easy to use
  • Accurate
Hach Method 10129. Range: 0.3 - 20.0 mg/L C. Set includes digestion vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed). Requires digestion.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.